Learn More
Medchemexpress LLC Mal-PEG8-Val-Cit-PAB-MMAE | 3058970-85-4 | 97.1% | 1626.97 | 1 MG
SDP
Supplier:  Medchemexpress LLC 1MGHY141156
Encompass_Preferred
Mal-PEG8-Val-Cit-PAB-MMAE is an agent-linker conjugate designed for Antibody-Drug Conjugates (ADCs). This compound incorporates a cleavable ADC linker and the potent tubulin inhibitor MMAE. ADCs consist of an antibody to which an ADC cytotoxin is attached via an ADC linker. It is for research use only and has not been fully validated for medical applications.
  • Contains a cleavable ADC linker.
  • Includes MMAE, a potent tubulin inhibitor.
  • Suitable for Antibody-Drug Conjugate (ADC) research.
Catalog No. 50-003-65469