Learn More
Medchemexpress LLC Mal-PEG8-val-cit-pab-mmaf | 1640.95 | C81H129N11O24 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY1567581MG
Encompass_Preferred
Mal-PEG8-Val-Cit-PAB-MMAF is a drug-linker conjugate for antibody-drug conjugate (ADC) research that links a maleimide-PEG8 spacer to a Val-Cit-PAB cleavable linker and a tubulin inhibitor payload to enable targeted intracellular release.
  • Cleavable Val-Cit-PAB linker for protease-triggered release.
  • Maleimide group for thiol conjugation to antibodies.
  • PEG8 spacer to increase hydrophilicity and solubility.
  • High molecular weight (1640.95) and defined molecular formula.
  • Soluble in DMSO (reported 100 mg/mL).
  • Intended for research use only.
Catalog No. 50-002-04432