Learn More
Medchemexpress LLC Mal-Ph-CONH-PEG4-NHS ester | 1263044-88-7 | 561.54 | 50 MG
SDP
Supplier:  Medchemexpress LLC 50MGHY133545
Encompass_Preferred
Mal-Ph-CONH-PEG4-NHS ester is a non-cleavable 4 unit PEG ADC linker. It is utilized in the synthesis of antibody-drug conjugates (ADCs), which consist of an antibody attached to an ADC cytotoxin via an ADC linker.
  • Non-cleavable 4 unit PEG ADC linker
  • Used in the synthesis of antibody-drug conjugates (ADCs)
  • Applicable as laboratory chemicals
  • Suitable for the manufacture of substances
Catalog No. 50-003-64110