Learn More
Medchemexpress LLC Malachite green carbinol base | 510-13-4 | 92.2% | 346.47 g/mol | C23H26N2O | 10 MG
SDP
Supplier:  Medchemexpress LLC HYD120010MG
Encompass_Preferred
Malachite Green Carbinol base is a leucohydroxide derivative of malachite green supplied as a solid for analytical, staining, and dyeing applications. It responds to UV irradiation with a rapid color change and can be used in pH-regulation experiments, textile dyeing, and biological staining or parasiticide studies.
  • Leucohydroxide derivative suitable for analytical use.
  • Rapid color change under UV irradiation enabling pH-regulation studies.
  • Compatible with dyeing of textiles and laboratory staining.
  • Used as a fungicide/parasiticide in aquaculture research.
  • Provided in small pack sizes for analytical workflows.
Catalog No. 50-002-97135