Learn More
Medchemexpress LLC Malachite Green Carbinol base | 510-13-4 | MFCD00151208 | 92.2% | 346.47 | 50 MG

Supplier: Medchemexpress LLC HYD120050MG
Malachite Green Carbinol base, also known as bis[4-(dimethylamino)phenyl](phenyl)methanol, is a chemical compound with the molecular formula C23H26N2O and a molecular weight of 346.47. It is typically supplied as a green to dark green solid.
- Molecular formula: C23H26N2O
- Molecular weight: 346.47
- Purity (LCMS): 92.2%
- Appearance: Green to dark green solid
- Storage: 4°C, protect from light. In solvent: -80°C, 6 months; -20°C, 1 month (protect from light).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.