Learn More
Medchemexpress LLC Malachite green-d5 oxalate | 423.52 g/mol | C25H21D5N2O4 | 2.5 MG
SDP
Supplier:  Medchemexpress LLC HYW6542542.5MG
Encompass_Preferred
Malachite green-d5 oxalate is the deuterium-labeled isotopologue of malachite green provided as the oxalate salt for research and analytical use. It is intended for use as a tracer or internal standard in mass spectrometry and other analytical workflows, supplied in a small research-scale quantity.
  • Deuterium-labeled internal standard for quantitative analysis.
  • Provided as an oxalate salt for enhanced stability and handling.
  • Suitable for method development and trace-level quantification in analytical assays.
  • Certificate of analysis and spectral data are available on request to confirm purity.
  • Store and handle according to the certificate of analysis and safety data sheet.
Catalog No. 50-002-83920