Learn More
Apexbio Technology LLC Malachite Green Phosphate Assay Kit 1000T
SDP
Supplier:  Apexbio Technology LLC K22451000
Encompass_Preferred
Malachite Green Phosphate Assay Kit is a colorimetric assay kit designed to quantify free inorganic phosphate in biological samples It measures phosphate concentration by forming a phosphomolybdate complex under acidic conditions producing a detectable absorption peak at approximately 650 nm The assay is primarily used to assess phosphate release in enzyme-activity assays such as ATPase reactions and other phosphate-releasing enzymatic studies Based on these properties Malachite Green Phosphate Assay Kit holds research potential in enzymology biochemical assays and phosphate metabolism analysis
Catalog No. 50-000-97128