Learn More
Sigma Aldrich Fine Chemicals Biosciences Malate Dehydrogenase Assay
Supplier:  Sigma Aldrich Fine Chemicals Biosciences MAK1961KT
Encompass
Malate dehydrogenase (MDH EC 1.1.1.37) is an NAD oxidoreductase that catalyzes the NAD/NADH-dependent interconversion of malate and oxalacetate. This reaction is critical to maintain the malate/aspartate shuttle across the mitochondrial membrane and Krebs tricarboxylic acid cycle in the mitochondrial matrix. Increased MDH activity has been observed in neurodegenerative diseases such as Alzheimers disease.
Catalog No. 50-290-4356