Learn More
Cayman Chemical MalathIon-d10 10mg
SDP
Supplier:  Cayman Chemical 3956110 MG
Encompass_Preferred
An internal standard for the quantification of malathion by GC- or LC-MS
Catalog No. 50-291-8489