Learn More
New Jersey Fluid Systems LLC MALE CONNECTOR SS-100-1-4
Supplier:  New Jersey Fluid Systems LLC SS10014
Encompass
316 Stainless Steel Swagelok Tube Fitting Male Connector. 1/16 in. OD x 1/4 in. male NPT
Catalog No. NC0198379