Learn More
Foxx Life Sciences CPC MPC Connector Coupling Insert (Male) MPC22006T03M - MPC to 3/8"" HB, Non-Valved, Polycarbonate (PC), Class VI, 50/PK
Supplier:  Foxx Life Sciences 2581320FLS
Encompass_Preferred
CPC MPC Couplings, MPC22006T03M, provide a critical function through sterile connections, even in non-sterile environments. Ease of use design allows for quick connections and the ability to transfer media easily with less risk of error. Reliable and repeatable performance eliminates the need for clamps, fixtures or other additional components for high quality single-use connections every time. security to your most critical fluid handling applications. CPC MPC Couplings offer optional locking sleeves to further guard against accidental disconnects. In addition, coupling halves can be rotated when connected to reduce tube kinks. They are reusable, yet economical enough to allow disposability. Fitting materials pass all physicochemical, cytotoxicity, leachables, and extractables tests. Choose from a full line of connectors and configurations, including pressure sealing caps and plugs, available in sizes 1/8", 1/4"&3/8".USP Class VI CertifiedADCF ComplaintBSE/TSE free materials
Catalog No. NC1795957