Learn More
Cole-Parmer Idex Flangeless Male Nut, FlushNut™, SS, 1/8" OD Tubing, 1/4-28 Flat-Bottom; 1/EA
Supplier:  Cole-Parmer 0201922
Encompass
Just slip the nut and ferrule over the tubing and finger tighten the assembly into your receiving port. Unlike the time required to flange tubing, it only takes a few seconds to change the components. Select from various ferrule materials to suit your application.
Catalog No. NC2052804