Learn More
Sigma Aldrich Fine Chemicals Biosciences Maleimide Activated KLH5MG
Supplier:  Sigma Aldrich Fine Chemicals Biosciences K03835MG
Encompass
Maleimide Activated KLH is provided in lyophilized ready to-use form eliminating the need for time-consuming activation and purification steps of the carrier protein. Maleimide Activated KLH contains approximately 160-320 maleimide groups for each KLH molecule. The conjugation reaction with a cysteine containing peptide is performed at pH 6.6-7.0. The extent of conjugation can then be determined by a colorimetric assay with Ellmans reagent to detect unreacted thiol groups in the cysteine-containing peptide.
Catalog No. 50-290-4335