Learn More
Medchemexpress LLC HY-133540 50mg Medchemexpress, Maleimide-DOTA CAS:1006711-90-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13354050MG
Encompass_Preferred
Medchemexpress, HY-133540 50mg Maleimide-DOTA CAS:1006711-90-5 Maleimide-DOTA is a non-cleavable ADC linker used in the synthesis of antibody-drug conjugates (ADCs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0295
 Restricted Item