Learn More
Medchemexpress LLC Maleimide-nota | 1295584-83-6 | MFCD32689663 | 99.3% | 425.44 g/mol | C18H27N5O7 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHYW087027
Encompass_Preferred
Maleimide-NOTA is a bifunctional NOTA-derived chelator for labeling peptides and antibodies via cysteine conjugation. It contains a thiol-reactive maleimide group for selective reaction with cysteine residues and a NOTA macrocycle for chelation of metal ions used in radiolabeling and imaging applications.
  • thiol-selective maleimide functional group for cysteine conjugation.
  • NOTA macrocycle suitable for radiometal and metal chelation.
  • reported high purity in supplier documentation.
  • available in small research pack sizes, including 25 mg.
  • use for peptide and antibody labeling in preclinical and imaging research.
Catalog No. 50-002-08900