Learn More
Medchemexpress LLC Maleimide-PEG-COOH | 96.8% | 2000 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY174921A100MG
Encompass_Preferred
Maleimide-PEG-COOH (MW 2000) is a PEG derivative composed of Maleimide, PEG units, and carboxyl (-COOH). The carboxyl group can readily form a stable amide bond with amino groups or an ester bond with hydroxyl groups. Maleimide forms a stable thioether bond with sulfhydryl (-SH).
  • A PEG derivative composed of maleimide, PEG units, and carboxyl (-COOH)
  • Carboxyl can form a stable amide bond with amino groups or an ester bond with hydroxyl groups
  • Maleimide forms a stable thioether bond with sulfhydryl (-SH)
Catalog No. 50-003-87086