Learn More
Medchemexpress LLC Maleimide-PEG-COOH | 96.8% | 2000 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY174921A500MG
Encompass_Preferred
Maleimide-PEG-COOH (MW 2000) is a PEG derivative consisting of Maleimide, PEG units, and a carboxyl (-COOH) group. The carboxyl group readily forms a stable amide bond with amino groups or an ester bond with hydroxyl groups. The Maleimide component forms a stable thioether bond with sulfhydryl (-SH) groups.
  • Carboxyl group forms a stable amide bond with amino groups or an ester bond with hydroxyl groups.
  • Maleimide forms a stable thioether bond with sulfhydryl (-SH) groups.
  • For research use only.
Catalog No. 50-003-87087