Learn More
Medchemexpress LLC Maleimide-PEG-COOH | 96.8% | 2000 (Average) | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY174921A
Encompass_Preferred
Maleimide-PEG-COOH (MW 2000) is a PEG derivative composed of Maleimide, PEG units, and carboxyl (-COOH). The carboxyl group can easily form a stable amide bond with amino groups or an ester bond with hydroxyl groups. Maleimide forms a stable thioether bond with sulfhydryl (-SH). This product is for research use only.
  • Peg derivative.
  • Carboxyl group forms stable amide bond with amino groups or ester bond with hydroxyl groups.
  • Maleimide forms stable thioether bond with sulfhydryl (-SH).
Catalog No. 50-003-85547