Learn More
LAYSAN BIO INC Maleimide-PEG-Succinimidyl Valarate (MAL-PEG-SVA) molecular weight: 5,000., 1gram
Supplier:  LAYSAN BIO INC MALPEGSVA50001G
Encompass
Maleimide-PEG-SVA (Succinimidyl Valerate) is a heterobifunctional combining a maleimide group at one end of the PEG chain and a succinimidyl valerate group at the other. This unique structure allows it to react with both thiol (-SH) and amine (-NH2) groups, making it highly versatile for various bioconjugation applications.
Catalog No. NC3876845