Learn More
Sigma Aldrich Fine Chemicals Biosciences Maleimide PEG5K OH average1G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences JKA5036
Encompass
Maleimide PEG Hydroxyl. Maleimide is thiol reactive and Hydroxyl is stable.
Catalog No. 50-290-7339