Learn More
Medchemexpress LLC Maleimide-PEG8-Val-Ala-PABC | 99.1% | 867.98 g/mol | C41H65N5O15 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY13225250MG
Encompass_Preferred
Mal-PEG8-Val-Ala-PABC is a cleavable Tesirine-type antibody-drug conjugate (ADC) linker containing a maleimide reactive group, a PEG8 spacer, and a Val-Ala dipeptide PABC self-immolative spacer. It is designed for thiol conjugation to antibodies and protease-triggered release of payloads in ADC applications.
  • Cleavable by cathepsin B and related proteases.
  • Maleimide group enables thiol conjugation to biomolecules.
  • PEG8 spacer improves solubility and reduces steric hindrance.
  • High purity: 99.1%.
  • Viscous liquid, colorless to light pink.
  • Storage: keep sealed at -20°C; in solvent store at -80°C (6 months) or -20°C (1 month).
Catalog No. 50-002-02372