Learn More
Aat Bioquest Amplite™ Colorimetric Maleimide Quantitation Kit
SDP
Supplier:  Aat Bioquest 5525
Encompass_Preferred
Maleimides can be directly assayed spectrophotometrically at 302 nm. However, the small extinction coefficient of 620 M-1cm-1 renders this assay insensitive, and the assay is further complicated by the protein absorbance at the same wavelength.
Catalog No. 50-195-3293