Learn More
Medchemexpress LLC Maleimido-mono-amide-DOTA | 1006711-90-5 | MFCD32708535 | 99.6% | 526.54 | C22H34N6O9 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY133540100MG
Encompass_Preferred
Maleimide-DOTA is a non-cleavable antibody-drug conjugate (ADC) linker that combines a maleimide reactive group for thiol conjugation with a DOTA chelator for metal coordination and radiolabeling. It is supplied as a powder for research use and shows high solubility in common solvents and buffered formulations.
  • Non-cleavable ADC linker suitable for antibody-drug conjugation
  • Maleimide group enables thiol-specific conjugation
  • DOTA chelator supports metal chelation and radiolabeling
  • High solubility in DMSO and water; ultrasonic may be required
  • Compatible with common in vivo vehicle systems at lower concentrations
  • Store powder sealed at -20°C; in solution store at -80°C up to 6 months
Catalog No. 50-002-03519