Learn More
Medchemexpress LLC Maleimidocaproyl-L-valine-L-citrulline-p-aminobenzyl alcohol p-nitrophenyl carbonate | 159857-81-5 | MFCD22200431 | 98.7% | 737.76 g/mol | C35H43N7O11 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY20336500MG
Encompass_Preferred
Mc-Val-Cit-PABC-PNP is a cathepsin-cleavable linker used in the synthesis of antibody-drug conjugates (ADCs). It couples an antibody-reactive maleimidocaproyl moiety to a valine-citrulline dipeptide cleavage site with a p-aminobenzyl alcohol self-immolative spacer and a p-nitrophenyl carbonate leaving group for payload attachment.
  • Cathepsin-cleavable valine-citrulline linker for controlled payload release.
  • Maleimidocaproyl functional group enables thiol-selective antibody conjugation.
  • P-aminobenzyl alcohol spacer supports self-immolation after cleavage.
  • P-nitrophenyl carbonate leaving group facilitates payload coupling.
  • High purity suitable for research-grade ADC synthesis.
  • Available as a 500 MG package for preparative work.
Catalog No. 50-002-02652