Learn More
Medchemexpress LLC Maleimidocaproyl-monomethylauristatin E | 863971-24-8 | MFCD28167757 | 99.9% | 911.18 | C49H78N6O10 | 25 MG
SDP
Supplier:  Medchemexpress LLC HY1574125MG
Encompass_Preferred
Mc-MMAE (maleimidocaproyl-monomethylauristatin E) is a maleimidocaproyl-protected monomethyl auristatin E used as an agent-linker conjugate in antibody-drug conjugate research. It functions as a potent tubulin inhibitor and is supplied as a solid for conjugation and synthesis of ADCs.
  • Protective maleimidocaproyl linker conjugated to monomethyl auristatin E.
  • Potent tubulin inhibitor for ADC payload development.
  • High purity suitable for research applications.
  • Solid form, white to light yellow, unstable in solution; prepare fresh.
  • Molecular formula C49H78N6O10 and molecular weight 911.18.
Catalog No. 50-002-04504