Learn More
Medchemexpress LLC Maleimidocaproyl monomethylauristatin F | 863971-19-1 | MFCD25976741 | >99.0% | 925.2 g/mol | C49H76N6O11 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY1557850MG
Encompass_Preferred
McMMAF (maleimidocaproyl monomethylauristatin F) is a maleimidocaproyl-protected derivative of monomethylauristatin F used as a cytotoxic payload and linker reagent in antibody-drug conjugate (ADC) development. It contains a thiol-reactive maleimide for conjugation to antibodies and delivers a potent tubulin polymerization inhibitor to target cells.
  • Maleimidocaproyl-conjugated monomethylauristatin F for ADC applications.
  • Thiol-reactive maleimide enables site-selective antibody conjugation.
  • Potent tubulin inhibitor payload for targeted cytotoxicity.
  • High purity suitable for research use; soluble in DMSO.
  • Store at -20°C to maintain stability.
Catalog No. 50-002-04425