Learn More
Apexbio Technology LLC Thiomyristoyl 1429749-41-6 5mg
SDP
Supplier:  Apexbio Technology LLC B79395
Encompass_Preferred
Thiomyristoyl (CAS 1429749-41-6) is a selective inhibitor of SIRT2 a NAD -dependent deacetylase involved in cell cycle regulation apoptosis metabolic processes and neurodegenerative disease mechanisms By binding specifically to SIRT2 Thiomyristoyl effectively suppresses its enzymatic activity displaying limited activity against other sirtuin family members In vitro studies report an IC50 of approximately 28 nM for SIRT2 inhibition Thiomyristoyl is widely employed as a chemical probe in cellular biology and signal transduction research facilitating investigation of SIRT2-related pathways and supporting the exploration of its biological functions and therapeutic potential
Catalog No. 50-000-87997