Learn More
Cusabio Technology LLC Mouse malonyl coenzyme A ELISA kit
Supplier:  Cusabio Technology LLC CSBE12896M
Encompass_Preferred
This Mouse malonyl CoA ELISA Kit was designed for the quantitative measurement of Mouse malonyl CoA protein in serum, plasma, tissue homogenates. It is a Sandwich ELISA kit, its detection range is 0.156 ng/mL-10 ng/mL and the sensitivity is 0.039 ng/mL. One kit contains 96 wells.
Catalog No. NC2952945