Learn More
Medchemexpress LLC Malonyl coenzyme A lithium | 108347-84-8 | MFCD00057769 | 98.0% | 860.6 g/mol | C24H38LiN7O19P3S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1364085MG
Encompass_Preferred
Malonyl CoA lithium is the lithium salt form of malonyl coenzyme A used as a biochemical reagent to modulate mitochondrial fatty acid metabolism. It functions as an inhibitor of carnitine palmitoyl transferase 1 (CPT1) and is intended for research use in studies of lipid metabolism and energy homeostasis.
  • Inhibitor of carnitine palmitoyl transferase 1 (CPT1).
  • Modulates fatty acid oxidation in cellular and biochemical assays.
  • Provided as the lithium salt to aid solubility in aqueous systems.
  • Useful for studies of mitochondrial metabolism and lipid regulation.
  • Accompanied by datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-21832