Learn More
Medchemexpress LLC Maltobionic acid (4-O-α-D-glucopyranosyl-D-gluconic acid) | 534-42-9 | MFCD30738187 | 99.9% | 358.30 | C12H22O12 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYW423191100MG
Encompass_Preferred
Maltobionic acid is a naturally derived polyhydroxy bionic acid used in biochemical and cellular research. It exhibits iron-chelating activity, antibacterial potential, and cytoprotective properties, and is supplied as a solid reagent suitable for analytical and biological studies.
  • Exhibits iron-chelating activity.
  • Shows antibacterial potential.
  • Provides cytoprotection in cellular studies.
  • Supplied as a white to off-white solid in multiple pack sizes.
  • Chemical formula C12H22O12; molecular weight 358.30.
  • High-purity reagent; reported purity values may vary by lot.
Catalog No. 50-002-81652