Learn More
Medchemexpress LLC Maltobionic acid (4-O-α-D-glucopyranosyl-D-gluconic acid) | 534-42-9 | MFCD30738187 | 99.9% | 358.30 g/mol | C12H22O12 | 1 ML

Supplier: Medchemexpress LLC HYW42319110MM/1ML
Maltobionic acid (4-O-α-D-glucopyranosyl-D-gluconic acid) is a naturally derived polyhydroxy bionic acid with reported iron-chelating activity, antibacterial potential, and cytoprotective properties. It is available as a powder and as a ready-to-use 10 mM solution in DMSO; manufacturer data include solubility and storage recommendations for in vitro and in vivo applications.
- High purity (99.86%).
- Water-soluble powder with recommended DMSO solubility for in vitro use.
- Provided as a 10 mM solution in DMSO for ready-to-use applications.
- Solubility protocols suitable for in vivo formulations (SBE-β-CD/saline or corn oil).
- Recommended storage conditions for powder and in-solution stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.