Learn More
Medchemexpress LLC Maltobionic acid | 534-42-9 | MFCD30738187 | 90.0% | 358.30 g·mol⁻¹ | C12H22O12 | 25 MG
SDP
Supplier:  Medchemexpress LLC HYW42319125MG
Encompass_Preferred
Maltobionic acid is a naturally derived polyhydroxy bionic acid used as a laboratory research reagent. It demonstrates iron-chelating activity, antibacterial potential, and cytoprotective effects, and is offered in both solid and solution formats for bench-scale studies. Intended for research use only and not for clinical or human use.
  • Exhibits iron-chelating ability.
  • Shows antibacterial potential.
  • Provides cytoprotective properties in studies.
  • Available in solid and DMSO solution formats.
  • Supplied in small research pack sizes suitable for screening.
  • Typical purity reported at 90.0%; batch purities may vary.
Catalog No. 50-002-80724