Learn More
Medchemexpress LLC Maltobionic acid | 534-42-9 | MFCD30738187 | 99.9% | 358.30 | C12H22O12 | 50 MG
SDP
Supplier:  Medchemexpress LLC HYW42319150MG
Encompass_Preferred
Maltobionic acid is a naturally derived polyhydroxy bionic acid with reported iron-chelating, antibacterial, and cytoprotective activities. It is supplied in solid and solution formats for laboratory use and formulation development.
  • Naturally derived polyhydroxy bionic acid.
  • Exhibits iron-chelating ability.
  • Shows antibacterial potential and cytoprotection.
  • Available as solid and solution formulations for flexible use.
  • Suitable for biochemical research and formulation studies.
  • Stable under recommended storage conditions when handled per safety data guidance.
Catalog No. 50-002-81653