Learn More
Medchemexpress LLC Maltol | 118-71-8 | ≥99.0% | 126.11 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01278810G
Encompass_Preferred
Maltol is an aromatic compound and an antioxidant agent. It enhances neural function by mitigating oxidative stress and cell apoptosis. It acts as an inhibitor of oxidative damage in nerve cells and is effective in preventing diabetic peripheral neuropathy (DPN). It is widely used as a safe flavoring agent and food preservative, and also functions as a metal ion chelator applicable in catalysis, cosmetics, and medicine.
  • Aromatic compound
  • Antioxidant agent
  • Enhances neural function
  • Mitigates oxidative stress and cell apoptosis
  • Inhibitor of oxidative damage in nerve cells
  • Effective in preventing diabetic peripheral neuropathy (DPN)
  • Used as a safe flavoring agent and food preservative
  • Metal ion chelator
  • Applicable in catalysis, cosmetics, and medicine
Catalog No. 50-004-54862