Learn More
Medchemexpress LLC Maltol | 118-71-8 | 99.98% | 126.11 | 50 G
SDP
Supplier:  Medchemexpress LLC HYW01278850G
Encompass_Preferred
Maltol, an aromatic compound, functions as an antioxidant agent. It improves neural function by reducing oxidative stress and cell apoptosis. This compound acts as an inhibitor of oxidative damage in nerve cells, proving effective in the prevention of diabetic peripheral neuropathy (DPN). Maltol is widely employed as a safe flavoring agent and food preservative. Additionally, it serves as a metal ion chelator, finding applications in catalysis, cosmetics, and medicine.
  • Antioxidant agent
  • Enhances neural function by mitigating oxidative stress and cell apoptosis
  • Inhibitor of oxidative damage in nerve cells
  • Effective in preventing diabetic peripheral neuropathy (DPN)
  • Used extensively as a safe flavoring agent and food preservative
  • Metal ion chelator applicable in catalysis, cosmetics, and medicine
Catalog No. 50-004-54836