Learn More
Carbosynth LLC. Maltooctaose, 66567-45-1, MFCD05865015, 100 mg
SDP
Supplier:  Carbosynth LLC. OM06941100MG
Encompass_Preferred
Chemical Formula: C48H82O41, MW: 1315.16. α 1,4-glucooctasaccharide derived from starch by hydrolysis and chromatography.
Catalog No. NC1106492