Learn More
Medchemexpress LLC HY-N2024 500mg Medchemexpress, Maltose CAS:69-79-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN2024500MG
Encompass_Preferred
Medchemexpress, HY-N2024 500mg Maltose CAS:69-79-4 Maltose is a disaccharide formed from two units of glucose joined with an α(1→4) bond. Maltose is a reducing sugar. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1472
 Restricted Item