Learn More
Pfanstiehl Inc Maltose Monohydrate. High Purity, Low Endotoxin, Low Metals, USP-NF, JP, ChP (M-167) -100G GMP, Multi-compendial CAS: 6363-53-7
Supplier:  Pfanstiehl Inc M167RD
Encompass
Pfanstiehl Maltose Monohydrate High Purity, Low Endotoxin, Low Metals is a multi-compendial carbohydrate utilized in bioprocessing applications and as an infusion stabilizer. Pfanstiehl Maltose has been specifically designed and formulated to remove impurities such as endotoxin, trace metals, and nitrosamines. Manufactured in the USA under ICH-Q7 cGMP conditions, Pfanstiehl Maltose meets parenteral (injectable) excipient grade standards, making it ideal as a bioprocessing feed, or as a stabilizing agent for pharmaceutical infusions. Pfanstiehl Maltose is available as USP-NF (National Formulary) JP (Japanese Pharmacopeia) and ChP (Chinese Pharmacopeia) Compendial, U.S.P. , cGMP , pharmaceutical grade .
Catalog No. NC2953567