Learn More
Medchemexpress LLC Malvidin-3-galactoside (chloride) | 30113-37-2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN66235MG
Encompass_Preferred
Malvidin-3-galactoside chloride is an anthocyanin monomer known for its anti-tumor function. It induces hepatocellular carcinoma (HCC) cell cycle arrest and apoptosis, while also inhibiting the production and accumulation of reactive oxygen species (ROS). This product is intended for research use only.
  • Induces hepatocellular carcinoma cell cycle arrest and apoptosis
  • Inhibits production and accumulation of reactive oxygen species
  • Displays cytotoxicity on Huh-7 cells
  • Increases levels of caspase-3 and cleaved-PARP
  • Reduces phosphorylation ERK levels
  • Increases phosphorylation level of p38 and JNK
  • Inhibits migration and invasion in Huh-7 cells
Catalog No. 50-004-18755