Learn More
Medchemexpress LLC Malvidin-3-glucoside chloride | 7228-78-6 | >95.0% | 528.89 | C23H25ClO12 | 1MG
SDP
Supplier:  Medchemexpress LLC HY1257401MG
Encompass_Preferred
Malvidin-3-glucoside chloride is an anthocyanin analytical standard used as a reference material in qualitative and quantitative analytical assays. It is applied in chromatographic and mass spectrometric methods and is investigated for anti-inflammatory and antioxidant effects related to NF-κB signaling and endothelial function. The compound is a solid with formula C23H25ClO12 and molecular weight 528.89; recommended storage is sealed, protected from moisture and light at 4°C, with solutions stored at -80°C (6 months) or -20°C (1 month).
  • Analytical standard for HPLC, GC, and MS applications.
  • Supports qualitative and quantitative assay development and validation.
  • Relevant for studies of anti-inflammatory and antioxidant activity.
  • Supplied in small pack sizes suitable for reference use.
  • Store sealed and protected from moisture and light; follow low-temperature storage for solutions.
Catalog No. 50-002-37843