Learn More
Medchemexpress LLC Malvidin-3-glucoside chloride | 7228-78-6 | 99.9% | 528.89 | C23H25ClO12 | 5MG
SDP
Supplier:  Medchemexpress LLC HY1257405MG
Encompass_Preferred
Malvidin-3-glucoside chloride is the chloride salt of the anthocyanin malvidin-3-O-glucoside. Supplied as a high-purity analytical standard, it is used in research on inflammation, oxidative stress, endothelial function, and gut microecology. The compound modulates NF-κB signaling and reduces expression of pro-inflammatory markers in cellular and animal studies.
  • High purity analytical standard for research use.
  • Modulates NF-κB signaling and inflammatory responses.
  • Exhibits antioxidant activity and endothelial protection.
  • Suitable for studies of intestinal microecology and vascular inflammation.
  • Available in mg-scale quantities for analytical assays.
Catalog No. 50-002-38137