Learn More
Cayman Chemical MAM2201 N 3fluoropentyl Is
SDP
Supplier:  Cayman Chemical 11781100 UG
Encompass_Preferred
Differs from MAM2201 by having fluorine at the 3 position rather than the 5 position of the alkyl chain; intended for forensic and research applications
Catalog No. 50-295-1994