Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ATCC Mammalian Orthoreovirus 1, Strain Lang (ATCC VR-230) - Frozen Research Virus

Supplier:  ATCC VR230

Encompass

Mammalian orthoreovirus 1 (strain Lang) is a frozen viral culture supplied for virology research and propagation. The isolate was derived from a rectal swab of a healthy child in Ohio, U.S., with an origin year of 1953. Next-generation sequencing identified an additional orthoreovirus (orthoreovirus 3) in the sample. For passage, harvest cell cultures after complete involvement of the cell sheet; culture fluid contains a group-specific CF antigen, and mycoplasma testing was negative. Store at -70°C or colder.

  • Format: frozen.
  • Storage: -70°C or colder.
  • Strain designation: Lang.
  • Isolation source: rectal swab from a healthy child (Ohio, United States; 1953).
  • Propagation host: LLC-MK2 derivative cell line.
  • Culture fluid contains group-specific CF antigen.
  • Mycoplasma not detected.

Catalog No. NC2206195


May include imposed supplier surcharges.

Due to product restrictions, please to purchase or view availability for this product.

This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.