Learn More
ABclonal Technology MAN1C1 Rabbit pAb
SDP
Supplier:  ABclonal Technology A128371000UL
Encompass_Preferred
Predicted to enable mannosyl-oligosaccharide 1,2-alpha-mannosidase activity. Predicted to be involved in N-glycan processing. Located in extracellular exosome.
Catalog No. 50-001-15449