Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mandelic acid | 90-64-2 | MFCD00064250 | 99.9% | 152.15 g·mol⁻1 | C8H8O3 | 100 G
SDP

Supplier:  Medchemexpress LLC HYW015591100G

Encompass_Preferred

Mandelic acid is an aromatic alpha-hydroxy carboxylic acid used as a building block and analytical standard in pharmaceutical and fine-chemical research. Supplied as a powder with certificate of analysis and safety data available, it offers high purity and stable storage profiles for laboratory use.

  • Alpha-hydroxy carboxylic acid suitable for synthesis and analytical applications.
  • Declared purity 99.92% (HPLC) for reproducible results.
  • Molecular weight 152.15 g·mol⁻1 and formula C8H8O3 for identity confirmation.
  • Available as a powder with recommended storage at -20°C to maintain stability.
  • Certificate of analysis and SDS available to support quality control.

Catalog No. 50-003-00678


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.