Learn More
Medchemexpress LLC Mangafodipir trisodium | 140678-14-4 | MFCD01939723 | 99.7% | 757.32 g·mol⁻¹ | C22H27MnN4Na3O14P2 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYB099310MG
Encompass_Preferred
Mangafodipir trisodium is the trisodium salt of mangafodipir, a manganese(II)-containing hepatocellular-specific magnetic resonance imaging (MRI) contrast agent with reported antioxidant and chemoprotective activity. Supplied as a brown to reddish-brown solid, it is water-soluble and intended for research applications in hepatic imaging and oxidative stress studies.
  • Hepatocellular-specific contrast enhancement for liver MRI.
  • Demonstrates antioxidant and chemoprotective properties in research models.
  • High purity suitable for analytical and experimental use.
  • Readily soluble in water with ultrasonic assistance.
  • Stable when stored sealed at 4 °C (solid); colder storage recommended for solutions.
Catalog No. 50-002-98191