Learn More
Alkali Scientific Manganese Chloride 1M Solution, 100 Milliliters
SDP
Supplier:  Alkali Scientific ANG001CP
Encompass_Preferred
Manganese Chloride 1M Solution is a laboratory reagent used primarily for preparing solutions in chemical and biological experiments. The 1M concentration offers a high level of manganese, providing a stable and reliable source for various analytical applications. It is commonly used in redox reactions, catalyst synthesis, and in biological assays where manganese ions play a crucial role in enzyme reactions. The 100mL volume is convenient for smaller laboratory setups and experiments. This reagent is essential for processes that require precise manganese concentration, including the study of metal ion interactions and the preparation of growth media for microorganisms in research settings.
Catalog No. 50-006-61087