Learn More
Strem, An Ascensus Company CAS# 7783-53-1. 5g. Manganese(III) fluoride, 98%. MFCD00016223
SDP
Supplier:  Strem, An Ascensus Company 9325425G
Encompass
CAS# 7783-53-1. 5g. Manganese(III) fluoride, 98%. MFCD00016223. Molecular Weight: 111.93. Molecular Formula: MnF3. Color/form: red pwdr. Strem# 93-2542. http://www.strem.com/catalog/v/93-2542/
Catalog No. 50-901-14620