Learn More
Strem, An Ascensus Company CAS# 1313-13-9. 50g. Manganese(IV) oxide, activated. MFCD00003463
SDP
Supplier:  Strem, An Ascensus Company 25136050G
Encompass
CAS# 1313-13-9. 50g. Manganese(IV) oxide, activated. MFCD00003463. Molecular Weight: 86.94. Molecular Formula: MnO2. Color/form: brown to black pwdr. Strem# 25-1360. http://www.strem.com/catalog/v/25-1360/
Catalog No. 50-901-10914