Learn More
Medchemexpress LLC Manganese peroxidase | 114995-15-2 | MFCD03792385 | 39,557 Da (≈39.6 kDa) | Not applicable - enzyme/protein (no single chemical formula) | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1321885MG
Encompass_Preferred
Manganese peroxidase is a heme-containing oxidative enzyme that catalyzes the oxidation of Mn2+ to Mn3+. It participates in lignin depolymerization and is used in laboratory research on biodegradation and environmental remediation, including dye wastewater decolorization. Supplied as a lyophilized powder for research use.
  • Heme-containing enzyme that oxidizes Mn2+ to Mn3+.
  • Facilitates lignin depolymerization for biomass and paper research.
  • Applicable to biodegradation and environmental contaminant removal.
  • Useful for dye wastewater decolorization and remediation studies.
  • Provided as lyophilized powder suitable for storage and reconstitution.
Catalog No. 50-002-62200